2-Fluorophenylboronic acid, 98%
2-Fluorophenylboronic acid, 98%
2-Fluorophenylboronic acid, 98%
Thermo Scientific Chemicals

2-Fluorophenylboronic acid, 98%

CAS: 1993-03-9 | C6H6BFO2 | 139.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 359340050
Price (EUR)
142,00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
142,00
Each
Chemical Identifiers
CAS1993-03-9
IUPAC Name(2-fluorophenyl)boronic acid
Molecular FormulaC6H6BFO2
InChI KeyQCSLIRFWJPOENV-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only