4-Iodophenylboronic acid, 97%
4-Iodophenylboronic acid, 97%
4-Iodophenylboronic acid, 97%
Thermo Scientific Chemicals

4-Iodophenylboronic acid, 97%

CAS: 5122-99-6 | C6H6BIO2 | 247.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 359480250
Price (EUR)
326,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
326,00
Each
Chemical Identifiers
CAS5122-99-6
IUPAC Name(4-iodophenyl)boronic acid
Molecular FormulaC6H6BIO2
InChI KeyPELJYVULHLKXFF-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Light beige
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only