6-Chloro-5-nitro-2-picoline, 98%
6-Chloro-5-nitro-2-picoline, 98%
6-Chloro-5-nitro-2-picoline, 98%
Thermo Scientific Chemicals

6-Chloro-5-nitro-2-picoline, 98%

CAS: 56057-19-3 | C6H5ClN2O2 | 172.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 360830050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56057-19-3
IUPAC Name2-chloro-6-methyl-3-nitropyridine
Molecular FormulaC6H5ClN2O2
InChI KeyUIEVSGOVFXWCIK-UHFFFAOYSA-N
SMILESCC1=CC=C(C(Cl)=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Yellow to brown
Melting point69°C to 74°C
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only