alpha-Bromostyrene, 95%, stabilized
alpha-Bromostyrene, 95%, stabilized
alpha-Bromostyrene, 95%, stabilized
Thermo Scientific Chemicals

alpha-Bromostyrene, 95%, stabilized

CAS: 98-81-7 | C8H7Br | 183.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 365800250
Price (EUR)
248,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
248,00
Each
Chemical Identifiers
CAS98-81-7
IUPAC Name(1-bromoethenyl)benzene
Molecular FormulaC8H7Br
InChI KeySRXJYTZCORKVNA-UHFFFAOYSA-N
SMILESBrC(=C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless to orange
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5870 to 1.5910 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only