N-(alpha,alpha,alpha-Trifluoro-p-tolyl)piperazine, 98%
N-(alpha,alpha,alpha-Trifluoro-p-tolyl)piperazine, 98%
N-(alpha,alpha,alpha-Trifluoro-p-tolyl)piperazine, 98%
Thermo Scientific Chemicals

N-(alpha,alpha,alpha-Trifluoro-p-tolyl)piperazine, 98%

CAS: 30459-17-7 | C11H13F3N3 | 230.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 367120010
Price (EUR)
47,50
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,50
Each
Chemical Identifiers
CAS30459-17-7
IUPAC Name1-[4-(trifluoromethyl)phenyl]piperazine
Molecular FormulaC11H13F3N2
InChI KeyIBQMAPSJLHRQPE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Crystals
Infrared spectrumConforms
GC>=97.5 %
Melting point85°C to 90°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only