2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%
Thermo Scientific Chemicals

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%

CAS: 191171-55-8 | C12H18BNO2 | 219.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 367560010
Price (EUR)
81,80
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
81,80
Each
Chemical Identifiers
CAS191171-55-8
IUPAC Name2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H18BNO2
InChI KeyZCJRWQDZPIIYLM-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point64°C to 70°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only