4,5-Dicyanoimidazole, 99%
4,5-Dicyanoimidazole, 99%
4,5-Dicyanoimidazole, 99%
Thermo Scientific Chemicals

4,5-Dicyanoimidazole, 99%

CAS: 1122-28-7 | C5H2N4 | 118.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 368530050
Price (EUR)
36,65
Online exclusive
40,30
Save 3,65 (9%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
36,65
Online exclusive
40,30
Save 3,65 (9%)
Each
Chemical Identifiers
CAS1122-28-7
IUPAC Name1H-imidazole-4,5-dicarbonitrile
Molecular FormulaC5H2N4
InChI KeyXGDRLCRGKUCBQL-UHFFFAOYSA-N
SMILESN#CC1=C(N=CN1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light beige
Infrared spectrumConforms
HPLC>=98.5 %
Melting point173°C to 177°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only