Indium(III) trifluoromethanesulfonate, 98%
Indium(III) trifluoromethanesulfonate, 98%
Indium(III) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Indium(III) trifluoromethanesulfonate, 98%

CAS: 128008-30-0 | C3F9InO9S3 | 562.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 368540050
Price (EUR)
96,65
Online Exclusive
107,00
Save 10,35 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
96,65
Online Exclusive
107,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS128008-30-0
IUPAC Nameindium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9InO9S3
InChI KeyUCYRAEIHXSVXPV-UHFFFAOYSA-K
SMILES[In+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric19.9 to 20.9 % (In)
Appearance (Color)White to almost white
Appearance (Form)Powder and/or granules
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only