Methylamine, pure, 2M solution in methanol, AcroSeal™
Thermo Scientific Chemicals

Methylamine, pure, 2M solution in methanol, AcroSeal™

CAS: 74-89-5 | CH5N | 31.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 368551000
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS74-89-5
IUPAC Namemethanamine
Molecular FormulaCH5N
InChI KeyBAVYZALUXZFZLV-UHFFFAOYSA-N
SMILESCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Strength2 M (typically) exact strength on the certificate of analysis
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only