4-Octylbenzaldehyde, 97%
4-Octylbenzaldehyde, 97%
4-Octylbenzaldehyde, 97%
4-Octylbenzaldehyde, 97%
Thermo Scientific Chemicals

4-Octylbenzaldehyde, 97%

CAS: 49763-66-8 | C15H22O | 218.34 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3688800505 gGlass bottle
Catalog number 368880050
Price (EUR)
119,85
Online Exclusive
141,00
Save 21,15 (15%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
119,85
Online Exclusive
141,00
Save 21,15 (15%)
Each
Add to cart
Chemical Identifiers
CAS49763-66-8
IUPAC Name4-octylbenzaldehyde
Molecular FormulaC15H22O
InChI KeyIHKVZPVHTKOSLW-UHFFFAOYSA-N
SMILESCCCCCCCCC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5100 to 1.5130 (20°C, 589 nm)
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only