Ethynylcyclopropane, 98%
Ethynylcyclopropane, 98%
Ethynylcyclopropane, 98%
Ethynylcyclopropane, 98%
Thermo Scientific Chemicals

Ethynylcyclopropane, 98%

CAS: 6746-94-7 | C5H6 | 66.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 369130100
Price (EUR)
82,40
Each
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,40
Each
Add to cart
Chemical Identifiers
CAS6746-94-7
IUPAC Nameethynylcyclopropane
Molecular FormulaC5H6
InChI KeyNPTDXPDGUHAFKC-UHFFFAOYSA-N
SMILESC#CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4270 to 1.4310 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only