tert-Butyl 2,2,2-trichloroacetimidate, 95%
tert-Butyl 2,2,2-trichloroacetimidate, 95%
tert-Butyl 2,2,2-trichloroacetimidate, 95%
Thermo Scientific Chemicals

tert-Butyl 2,2,2-trichloroacetimidate, 95%

CAS: 98946-18-0 | C6H10Cl3NO | 218.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 369330050
Price (EUR)
48,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
48,20
Each
Chemical Identifiers
CAS98946-18-0
IUPAC Nametert-butyl 2,2,2-trichloroethanimidate
Molecular FormulaC6H10Cl3NO
InChI KeyCQXDYHPBXDZWBA-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=N)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Appearance (Form)Liquid or low melting crystalline mass
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only