Platinum(II) acetylacetonate, 98%
Platinum(II) acetylacetonate, 98%
Platinum(II) acetylacetonate, 98%
Thermo Scientific Chemicals

Platinum(II) acetylacetonate, 98%

CAS: 15170-57-7 | C10H14O4Pt | 393.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 369350250
Price (EUR)
2.972,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
2.972,00
Each
Chemical Identifiers
CAS15170-57-7
IUPAC Nameplatinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O4Pt
InChI KeyKLFRPGNCEJNEKU-FDGPNNRMSA-L
SMILES[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97.5 % (ex Pt)
Appearance (Form)Powder
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only