Ethylenebis(triphenylphosphine)platinum(0), 98%
Ethylenebis(triphenylphosphine)platinum(0), 98%
Ethylenebis(triphenylphosphine)platinum(0), 98%
Thermo Scientific Chemicals

Ethylenebis(triphenylphosphine)platinum(0), 98%

CAS: 12120-15-9 | C38H34P2Pt | 747.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 369360010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12120-15-9
IUPAC Nameethene bis(triphenylphosphane) platinum
Molecular FormulaC38H34P2Pt
InChI KeyRZUASTIKPBCXPU-UHFFFAOYSA-N
SMILES[Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97.5 % (ex Pt)
Melting point127°C to 131°C
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only