Methyl thiosalicylate, 97%
Methyl thiosalicylate, 97%
Methyl thiosalicylate, 97%
Thermo Scientific Chemicals

Methyl thiosalicylate, 97%

CAS: 4892-02-8 | C8H8O2S | 168.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 369600050
Price (EUR)
82,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,20
Each
Chemical Identifiers
CAS4892-02-8
IUPAC Namemethyl 2-sulfanylbenzoate
Molecular FormulaC8H8O2S
InChI KeyBAQGCWNPCFABAY-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light green
Appearance (Form)Liquid
Refractive index1.5910 to 1.5950 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only