Eaton's reagent
Eaton's reagent
Eaton's reagent
Eaton's reagent
Thermo Scientific Chemicals

Eaton's reagent

CAS: 39394-84-8 | O10P4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Catalog number 369800025
Price (EUR)
436,00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (EUR)
436,00
Each
Chemical Identifiers
CAS39394-84-8, 75-75-2, 1314-56-3
Specifications
Boiling Point122.0°C (1.0 mmHg)
CAS39394-84-8
CAS75-75-2
CAS1314-56-3
Chemical Name or MaterialEaton's reagent
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only