3-Methoxycarbonylphenylboronic acid, 97%
3-Methoxycarbonylphenylboronic acid, 97%
3-Methoxycarbonylphenylboronic acid, 97%
Thermo Scientific Chemicals

3-Methoxycarbonylphenylboronic acid, 97%

CAS: 99769-19-4 | C8H9BO4 | 179.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 369830250
Price (EUR)
363,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
363,00
Each
Chemical Identifiers
CAS99769-19-4
IUPAC Name[3-(methoxycarbonyl)phenyl]boronic acid
Molecular FormulaC8H9BO4
InChI KeyALTLCJHSJMGSLT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=96 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only