Ethylamine, 2M solution in THF, AcroSeal™
Product Image
Thermo Scientific Chemicals

Ethylamine, 2M solution in THF, AcroSeal™

CAS: 75-04-7 | C2H7N | 45.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 370258000
Price (EUR)
-
Quantity:
800 mL
Request bulk or custom format
Chemical Identifiers
CAS75-04-7
IUPAC Nameethanamine
Molecular FormulaC2H7N
InChI KeyQUSNBJAOOMFDIB-UHFFFAOYSA-N
SMILESCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Strength1.90 to 2.20 M
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only