Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Thermo Scientific Chemicals

Sand, pure, 40-100 mesh

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 370940050
Price (EUR)
288,00
Each
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
288,00
Each
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle size207 µm (AGS) (typical)
Silicon dioxide (SiO2)99.77 % (typical)
Iron(III)oxide (Fe2O3)0.009 % (typical)
Aluminium oxide (Al2O3)0.030 % (typical)
Particle size207 µm (AGS) (typical)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only