L-Asparagine, 99%
L-Asparagine, 99%
L-Asparagine, 99%
Thermo Scientific Chemicals

L-Asparagine, 99%

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 371600025
Price (EUR)
1.401,00
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
1.401,00
Each
Chemical Identifiers
CAS70-47-3
IUPAC Name2-amino-3-carbamoylpropanoic acid
Molecular FormulaC4H8N2O3
InChI KeyDCXYFEDJOCDNAF-UHFFFAOYNA-N
SMILESNC(CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Water=<1 %
Specific optical rotation+33° to +36° (20°C, 589 nm) (c=10, 2N HCl)
Titration with HClO4>=98.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only