2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
Thermo Scientific Chemicals

2-Cyanobenzaldehyde, 98%

CAS: 7468-67-9 | C8H5NO | 131.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3718600505 gGlass bottle
Catalog number 371860050
Price (EUR)
75,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
75,00
Each
Chemical Identifiers
CAS7468-67-9
IUPAC Name2-formylbenzonitrile
Molecular FormulaC8H5NO
InChI KeyQVTPWONEVZJCCS-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to orange
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
GC>=97.5 %
Melting point102°C to 106°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only