Cyclopentyl isocyanide, 96%
Cyclopentyl isocyanide, 96%
Cyclopentyl isocyanide, 96%
Thermo Scientific Chemicals

Cyclopentyl isocyanide, 96%

CAS: 68498-54-4 | C6H9N | 95.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 372090010
Price (EUR)
151,65
Online exclusive
168,00
Save 16,35 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
151,65
Online exclusive
168,00
Save 16,35 (10%)
Each
Chemical Identifiers
CAS68498-54-4
IUPAC Nameisocyanocyclopentane
Molecular FormulaC6H9N
InChI KeyXTLSDEJNOQVCHL-UHFFFAOYSA-N
SMILES[C-]#[N+]C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=95 %
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only