2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%
2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%
2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%
Thermo Scientific Chemicals

2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%

CAS: 89992-70-1 | C9H18ClN2OP | 236.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 374830010
Price (EUR)
117,65
Online Exclusive
130,00
Save 12,35 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
117,65
Online Exclusive
130,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS89992-70-1
IUPAC Name3-({[bis(propan-2-yl)amino](chloro)phosphanyl}oxy)propanenitrile
Molecular FormulaC9H18ClN2OP
InChI KeyQWTBDIBOOIAZEF-UHFFFAOYNA-N
SMILESCC(C)N(C(C)C)P(Cl)OCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy colorless to yellow
Refractive index1.4760 to 1.4800 (20°C, 589 nm)
Chloride (Cl)14.5 to 15.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only