Dicyclohexyl phthalate, 99%
Dicyclohexyl phthalate, 99%
Dicyclohexyl phthalate, 99%
Thermo Scientific Chemicals

Dicyclohexyl phthalate, 99%

CAS: 84-61-7 | C20H26O4 | 330.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 375052500
Price (EUR)
49,20
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
49,20
Each
Chemical Identifiers
CAS84-61-7
IUPAC Name1,2-dicyclohexyl benzene-1,2-dicarboxylate
Molecular FormulaC20H26O4
InChI KeyVOWAEIGWURALJQ-UHFFFAOYSA-N
SMILESO=C(OC1CCCCC1)C1=CC=CC=C1C(=O)OC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White
Melting point63°C to 67°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only