3-Methylenecyclobutanecarbonitrile, 97%
3-Methylenecyclobutanecarbonitrile, 97%
3-Methylenecyclobutanecarbonitrile, 97%
Thermo Scientific Chemicals

3-Methylenecyclobutanecarbonitrile, 97%

CAS: 15760-35-7 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 376010050
Price (EUR)
79,70
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
79,70
Each
Chemical Identifiers
CAS15760-35-7
IUPAC Name3-methylidenecyclobutane-1-carbonitrile
Molecular FormulaC6H7N
InChI KeyZRWMAMOBIQQJSA-UHFFFAOYSA-N
SMILESC=C1CC(C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only