Titanium(IV) chloride, 1M solution in toluene, AcroSeal™
Titanium(IV) chloride, 1M solution in toluene, AcroSeal™
Titanium(IV) chloride, 1M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Titanium(IV) chloride, 1M solution in toluene, AcroSeal™

CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 378168000
Price (EUR)
243,65
Online Exclusive
271,00
Save 27,35 (10%)
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
243,65
Online Exclusive
271,00
Save 27,35 (10%)
Each
Chemical Identifiers
CAS7550-45-0
IUPAC Nametitanium(4+) tetrachloride
Molecular FormulaCl4Ti
InChI KeyXJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric0.95 to 1.1 M
Appearance (Color)Clear orange
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only