Propargyl p-toluenesulfonate, 97%
Propargyl p-toluenesulfonate, 97%
Propargyl p-toluenesulfonate, 97%
Thermo Scientific Chemicals

Propargyl p-toluenesulfonate, 97%

CAS: 6165-76-0 | C10H10O3S | 210.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 379680250
Price (EUR)
108,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
108,00
Each
Chemical Identifiers
CAS6165-76-0
IUPAC Nameprop-2-yn-1-yl 4-methylbenzene-1-sulfonate
Molecular FormulaC10H10O3S
InChI KeyLMBVCSFXFFROTA-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5270 to 1.5290 (20°C, 589 nm)
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only