2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 379721000
Price (EUR)
423,00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
423,00
Each
Add to cart
Chemical Identifiers
CAS5416-80-8
IUPAC Name2-methyl-1H-indole-3-carbaldehyde
Molecular FormulaC10H9NO
InChI KeyCYZIVXOEJNAIBS-UHFFFAOYSA-N
SMILESCC1=C(C=O)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.0 %
Melting point199°C to 202°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only