Product Image
Thermo Scientific Chemicals

Pheneturide

CAS: 90-49-3 | C11H14N2O2 | 206.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 381440050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS90-49-3
IUPAC Name(2-phenylbutanoyl)urea
Molecular FormulaC11H14N2O2
InChI KeyAJOQSQHYDOFIOX-UHFFFAOYNA-N
SMILESCCC(C(=O)NC(N)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals=<20 ppm
HPLC>=97.5 %
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only