Xylenes, 98%, pure, mixture of isomers
Xylenes, 98%, pure, mixture of isomers
Xylenes, 98%, pure, mixture of isomers
Xylenes, 98%, pure, mixture of isomers
Thermo Scientific Chemicals

Xylenes, 98%, pure, mixture of isomers

CAS: 1330-20-7 | C8H10
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
25 L
Packaging:
View all
Plastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 383930050
Price (EUR)
-
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=97 % (isomers and ethylbenzene)
Residue after evaporation=<0.01 %
Appearance (Form)Liquid
Water=<0.05 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only