4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%
Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%

CAS: 552846-17-0 | C14H23BN2O4 | 294.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 384640010
Price (EUR)
75,40
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
75,40
Each
Chemical Identifiers
CAS552846-17-0
IUPAC Nametert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
Molecular FormulaC14H23BN2O4
InChI KeyIPISOFJLWYBCAV-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point82°C to 89°C
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only