Glass Wool
Thermo Scientific Chemicals

Glass Wool

Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
50g
250g
1kg
Catalog number 386060500
Price (EUR)
21,65
Online exclusive
23,90
Save 2,25 (9%)
Each
Quantity:
50g
Request bulk or custom format
Price (EUR)
21,65
Online exclusive
23,90
Save 2,25 (9%)
Each
Specifications
Boiling Point1000°C
For Use With (Application)Filter aid
MaterialGlass
Melting Point680°C
Quantity50g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the diameter of Glass Wool (Cat. No. 386060010, 386060500, 386062500) threads?

Glass Wool (Cat. No. 386060010, 386060500, 386062500) threads have a diameter of 10-11 microns.