3,5-Di-tert-butylsalicylic acid, 99%
3,5-Di-tert-butylsalicylic acid, 99%
3,5-Di-tert-butylsalicylic acid, 99%
Thermo Scientific Chemicals

3,5-Di-tert-butylsalicylic acid, 99%

CAS: 19715-19-6 | C15H22O3 | 250.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 386860050
Price (EUR)
26,80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
26,80
Each
Chemical Identifiers
CAS19715-19-6
IUPAC Name3,5-di-tert-butyl-2-hydroxybenzoic acid
Molecular FormulaC15H22O3
InChI KeyZWQBZEFLFSFEOS-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(C(O)=O)=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point160°C to 165°C
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only