Petroleum ether, pure, boiling range ca. 180-280°C
Thermo Scientific Chemicals

Petroleum ether, pure, boiling range ca. 180-280°C

CAS: 8008-20-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 387490025
Price (EUR)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS8008-20-6
SpecificationsSpecification SheetSpecification Sheet
Density(15°C) >=0.79 kg/dm³
Aromatics=<0.2 %
Appearance (Form)Liquid
Boiling point200°C to 250°C
Refractive index1.4300 to 1.4500 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only