2-Hydroxyphenylboronic acid, 97%
2-Hydroxyphenylboronic acid, 97%
2-Hydroxyphenylboronic acid, 97%
Thermo Scientific Chemicals

2-Hydroxyphenylboronic acid, 97%

CAS: 89466-08-0 | C6H7BO3 | 137.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 387500050
Price (EUR)
197,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
197,00
Each
Chemical Identifiers
CAS89466-08-0
IUPAC Name(2-hydroxyphenyl)boronic acid
Molecular FormulaC6H7BO3
InChI KeyYDMRDHQUQIVWBE-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only