Thermo Scientific Chemicals

Acetyl chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 387741000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Acetyl chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 387741000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 387741000
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
75-36-5, 75-09-2
Appearance (Color)
Clear colorless
Titration Argentometric
0.9 to 1.1 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Carcinogenicity
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    H336 - May cause drowsiness or dizziness

    H351 - Suspected of causing cancer

    EU Specific Hazard Statements

    EUH014 - Reacts violently with water

    EUH071 - Corrosive to the respiratory tract

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician