Iron(II) bromide, 98%, anhydrous
Iron(II) bromide, 98%, anhydrous
Iron(II) bromide, 98%, anhydrous
Thermo Scientific Chemicals

Iron(II) bromide, 98%, anhydrous

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 388720500
Price (EUR)
321,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
321,00
Each
Chemical Identifiers
CAS7789-46-0
IUPAC Nameλ²-iron(2+) dibromide
Molecular FormulaBr2Fe
InChI KeyGYCHYNMREWYSKH-UHFFFAOYSA-L
SMILES[Fe++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to dark brown
Titration Argentometric>=97.5 %
Appearance (Form)Powder and/or lumps
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only