Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
Thermo Scientific Chemicals

Sodium, 99.8%, oiled sticks, wrapped in aluminum foil

CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 390121000
Price (EUR)
35,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
35,30
Each
Chemical Identifiers
CAS7440-23-5
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Sodium (Na)>=99.75 %
Potassium (K)=<300 ppm
Appearance (Form)Sticks
Calcium (Ca)=<400 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only