Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Thermo Scientific Chemicals

Dichloromethane, 99.5%, for analysis, stabilized with ethanol

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 390700010
Price (EUR)
42,65
Online exclusive
47,40
Save 4,75 (10%)
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
42,65
Online exclusive
47,40
Save 4,75 (10%)
Each
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.001 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only