1-Chloroisoquinoline, 97+%
1-Chloroisoquinoline, 97+%
1-Chloroisoquinoline, 97+%
Thermo Scientific Chemicals

1-Chloroisoquinoline, 97+%

CAS: 19493-44-8 | C9H6ClN | 163.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 391380050
Price (EUR)
107,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
107,00
Each
Chemical Identifiers
CAS19493-44-8
IUPAC Name1-chloroisoquinoline
Molecular FormulaC9H6ClN
InChI KeyMSQCQINLJMEVNJ-UHFFFAOYSA-N
SMILESClC1=C2C=CC=CC2=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.0 %
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Crystals and/or chunks or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only