Citric acid, trisodium salt, 98%, pure, anhydrous
Citric acid, trisodium salt, 98%, pure, anhydrous
Citric acid, trisodium salt, 98%, pure, anhydrous
Thermo Scientific Chemicals

Citric acid, trisodium salt, 98%, pure, anhydrous

CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 391970025
Price (EUR)
227,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
227,00
Each
Chemical Identifiers
CAS68-04-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only