3-Hydroxyphenylacetylene, 95+%
3-Hydroxyphenylacetylene, 95+%
3-Hydroxyphenylacetylene, 95+%
Thermo Scientific Chemicals

3-Hydroxyphenylacetylene, 95+%

CAS: 10401-11-3 | C8H6O | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 392410010
Price (EUR)
85,40
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
85,40
Each
Chemical Identifiers
CAS10401-11-3
IUPAC Name3-ethynylphenol
Molecular FormulaC8H6O
InChI KeyAODMJIOEGCBUQL-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95.0 %
Refractive index1.5840 to 1.5880 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only