4-Biphenylboronic acid, 97%
4-Biphenylboronic acid, 97%
4-Biphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Biphenylboronic acid, 97%

CAS: 5122-94-1 | C12H11BO2 | 198.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 394420050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5122-94-1
IUPAC Name{[1,1'-biphenyl]-4-yl}boronic acid
Molecular FormulaC12H11BO2
InChI KeyXPEIJWZLPWNNOK-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only