Dichlorobis(tricyclohexylphosphine)palladium(II), 98%
Dichlorobis(tricyclohexylphosphine)palladium(II), 98%
Dichlorobis(tricyclohexylphosphine)palladium(II), 98%
Thermo Scientific Chemicals

Dichlorobis(tricyclohexylphosphine)palladium(II), 98%

CAS: 29934-17-6 | C36H66Cl2P2Pd | 738.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 395892500
Price (EUR)
38,50
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
38,50
Each
Chemical Identifiers
CAS29934-17-6
IUPAC Namedichloropalladium; bis(tricyclohexylphosphane)
Molecular FormulaC36H66Cl2P2Pd
InChI KeyVUYVXCJTTQJVKJ-UHFFFAOYSA-L
SMILESCl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis>=97.5 %
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only