tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals
tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals
tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
396548000800mLAcroSeal™ Glass bottle
396541000100mLAcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 396548000
Price (EUR)
316,00
Each
Quantity:
800mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (EUR)
316,00
Each
Chemical Identifiers
CAS594-19-4
IUPAC Nametert-butyllithium
Molecular FormulaC4H9Li
InChI KeyBKDLGMUIXWPYGD-UHFFFAOYSA-N
SMILES[Li]C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Active base1.80 to 2.02 M
Appearance (Color)Clear to hazy colorless to yellow
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Active base17.0 to 19.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only