2,3-Pyrazinedicarboxylic anhydride, 97%
2,3-Pyrazinedicarboxylic anhydride, 97%
2,3-Pyrazinedicarboxylic anhydride, 97%
Thermo Scientific Chemicals

2,3-Pyrazinedicarboxylic anhydride, 97%

CAS: 4744-50-7 | C6H2N2O3 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 397130050
Price (EUR)
99,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
99,00
Each
Chemical Identifiers
CAS4744-50-7
IUPAC Name5H,7H-furo[3,4-b]pyrazine-5,7-dione
Molecular FormulaC6H2N2O3
InChI KeyAWJWCTOOIBYHON-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=NC=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration Acid>=96 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only