4-Bromo-2-iodoaniline, 97%
4-Bromo-2-iodoaniline, 97%
4-Bromo-2-iodoaniline, 97%
Thermo Scientific Chemicals

4-Bromo-2-iodoaniline, 97%

CAS: 66416-72-6 | C6H5BrIN | 297.91 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3979100505 gGlass bottle
Catalog number 397910050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS66416-72-6
IUPAC Name4-bromo-2-iodoaniline
Molecular FormulaC6H5BrIN
InChI KeyHHTYEQWCHQEJNV-UHFFFAOYSA-N
SMILESNC1=C(I)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to purple
Appearance (Form)Powder
Infrared spectrumConforms
Melting point68°C to 72°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only