mono-Methyl isophthalate, 97%
mono-Methyl isophthalate, 97%
mono-Methyl isophthalate, 97%
Thermo Scientific Chemicals

mono-Methyl isophthalate, 97%

CAS: 1877-71-0 | C9H8O4 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 398230010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1877-71-0
IUPAC Name3-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyWMZNGTSLFSJHMZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point191°C to 197°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only