Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%
Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%
Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%
Thermo Scientific Chemicals

Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%

CAS: 14337-43-0 | C4H6ClNO3 | 151.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 398740250
Price (EUR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14337-43-0
IUPAC Nameethyl (2Z)-2-chloro-2-(hydroxyimino)acetate
Molecular FormulaC4H6ClNO3
InChI KeyUXOLDCOJRAMLTQ-UTCJRWHESA-N
SMILESCCOC(=O)C(Cl)=NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point70°C to 75°C
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only