D-Mannosamine hydrochloride, 95%, Thermo Scientific Chemicals
D-Mannosamine hydrochloride, 95%, Thermo Scientific Chemicals
D-Mannosamine hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Mannosamine hydrochloride, 95%, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
3991700101 g
Catalog number 399170010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS5505-63-5
IUPAC Namehydrogen (2S,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanoic acid chloride
Molecular FormulaC6H14ClNO6
InChI KeyJMVMHPCPPSZGNY-BZWNWTOPSA-N
SMILES[H+].[Cl-].N[C@@H]([C@@H](O)[C@H](O)[C@H](O)CO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Specific optical rotation-2.5° to -6.5° (20°C, 589 nm) (C=1, water)
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only